PRIH8BA05A52 Princeton BioMolecular Research, Inc.

6-chloro-5-cyclopropylpyrazolo[1,5-a]pyrimidin-7-amine

  • MDL number MFCD31633877
  • Molecular Weight : 208.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart