PRIH8BA060AF Princeton BioMolecular Research, Inc.

3-methyl-5-(1-methyl-1H-pyrazol-4-yl)pyrazolo[1,5-a]pyrimidin-7-amine

  • MDL number MFCD31633881
  • Molecular Weight : 228.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart