PRIH8BA060B8 Princeton BioMolecular Research, Inc.

tert-butyl (3-amino-1-methyl-1H-pyrazol-4-yl)carbamate

  • MDL number MFCD31633890
  • Cas Number : 1773505-00-2
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart