PRIH8BA060C5 Princeton BioMolecular Research, Inc.

tert-butyl [1-(difluoromethyl)-3-nitro-1H-pyrazol-4-yl]carbamate

  • MDL number MFCD31633904
  • Molecular Weight : 278.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart