PRIH8BA060C8 Princeton BioMolecular Research, Inc.

1-methyl-3-nitro-1H-pyrazol-4-amine

  • MDL number MFCD00463970
  • Cas Number : 39205-76-0
  • Molecular Weight : 142.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart