PRIH8BA060CD Princeton BioMolecular Research, Inc.

1-(difluoromethyl)-3-nitro-1H-pyrazol-4-amine

  • MDL number MFCD31633912
  • Molecular Weight : 178.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart