PRIH8BA060DA Princeton BioMolecular Research, Inc.

tert-butyl (4-nitro-1-propyl-1H-pyrazol-3-yl)carbamate

  • MDL number MFCD31633924
  • Cas Number : 2023003-35-0
  • Molecular Weight : 270.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart