PRIH8BA060E6 Princeton BioMolecular Research, Inc.

tert-butyl [4-amino-1-(2-fluoroethyl)-1H-pyrazol-3-yl]carbamate

  • MDL number MFCD31633936
  • Molecular Weight : 244.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart