PRIH8BA060FC Princeton BioMolecular Research, Inc.

tert-butyl (4-amino-1-butyl-1H-pyrazol-5-yl)carbamate

  • MDL number MFCD31633961
  • Molecular Weight : 254.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart