PRIH8BA06104 Princeton BioMolecular Research, Inc.

tert-butyl (1-isobutyl-4-nitro-1H-pyrazol-5-yl)carbamate

  • MDL number MFCD31633969
  • Molecular Weight : 284.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart