PRIH8BA0610F Princeton BioMolecular Research, Inc.

1-sec-butyl-4-nitro-1H-pyrazol-5-amine

  • MDL number MFCD16093479
  • Cas Number : 1247716-36-4
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart