PRIH8BA06118 Princeton BioMolecular Research, Inc.

tert-butyl (5-amino-1-ethyl-1H-pyrazol-3-yl)carbamate

  • MDL number MFCD31633987
  • Molecular Weight : 226.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart