PRIH8BA0611A Princeton BioMolecular Research, Inc.

1-isopropyl-5-nitro-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD31633989
  • Molecular Weight : 199.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart