PRIH8BA06120 Princeton BioMolecular Research, Inc.

1-methyl-5-nitro-1H-pyrazole-4-carbonitrile

  • MDL number MFCD06166996
  • Cas Number : 91650-10-1
  • Molecular Weight : 152.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart