PRIH8BA06122 Princeton BioMolecular Research, Inc.

1-isopropyl-5-nitro-1H-pyrazole-4-carbonitrile

  • MDL number MFCD31633993
  • Molecular Weight : 180.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart