PRIH8BA06126 Princeton BioMolecular Research, Inc.

1-ethyl-3-nitro-1H-pyrazole-4-carbonitrile

  • MDL number MFCD31633997
  • Cas Number : 39205-93-1
  • Molecular Weight : 166.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart