PRIH8BA06132 Princeton BioMolecular Research, Inc.

3-amino-N-cyclopropyl-1-methyl-1H-pyrazole-4-carboxamide

  • MDL number MFCD31634007
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart