PRIH8BA0616E Princeton BioMolecular Research, Inc.

3-amino-1-isopropyl-N,N-dipropyl-1H-pyrazole-4-carboxamide

  • MDL number MFCD31634066
  • Molecular Weight : 252.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart