PRIH8BA0616F Princeton BioMolecular Research, Inc.

3-amino-1-(2,2-difluoroethyl)-N-methyl-1H-pyrazole-4-carboxamide

  • MDL number MFCD31634067
  • Molecular Weight : 204.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart