PRIH8BA06178 Princeton BioMolecular Research, Inc.

3-amino-1-(2,2-difluoroethyl)-N-(3-methoxypropyl)-1H-pyrazole-4-carboxamide

  • MDL number MFCD31634076
  • Molecular Weight : 262.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart