PRIH8BA06184 Princeton BioMolecular Research, Inc.

3-amino-N-methyl-1-(2,2,2-trifluoroethyl)-1H-pyrazole-4-carboxamide

  • MDL number MFCD31634088
  • Molecular Weight : 222.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart