PRIH8BA061A1 Princeton BioMolecular Research, Inc.

3-amino-1-cyclopentyl-N-(2-methoxyethyl)-1H-pyrazole-4-carboxamide

  • MDL number MFCD31634117
  • Molecular Weight : 252.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart