PRIH8BA061B3 Princeton BioMolecular Research, Inc.

3-amino-N-(sec-butyl)-1-(2-fluoroethyl)-1H-pyrazole-4-carboxamide

  • MDL number MFCD31634135
  • Molecular Weight : 228.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart