PRIH8BA061B8 Princeton BioMolecular Research, Inc.

3-amino-1-(2-fluoroethyl)-N,N-dimethyl-1H-pyrazole-4-carboxamide

  • MDL number MFCD31634140
  • Molecular Weight : 200.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart