PRIH93ED7A5C Princeton BioMolecular Research, Inc.

3-(methoxycarbonyl)-2,2,3-trimethylcyclopentanecarboxylic acid

  • MDL number MFCD00729046
  • Cas Number : 306935-16-0
  • Molecular Weight : 214.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart