PRIH93ED7A63 Princeton BioMolecular Research, Inc.

(2E)-7-(bromomethyl)-1,7-dimethylbicyclo[2.2.1]heptan-2-one oxime

  • MDL number MFCD00185726
  • Molecular Weight : 246.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart