PRIH93ED7A78 Princeton BioMolecular Research, Inc.

1-[2-(2-methylphenoxy)ethyl]-1H-benzimidazole

  • MDL number MFCD00839229
  • Molecular Weight : 252.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart