PRIH93ED7A89 Princeton BioMolecular Research, Inc.

(2-bromo-5-fluoro-1-benzofuran-3-yl)acetic acid

  • MDL number MFCD08690726
  • Molecular Weight : 273.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart