PRIH93ED7A9C Princeton BioMolecular Research, Inc.

(1H-Benzoimidazol-2-yl)-(4-fluoro-phenyl)-amine

  • MDL number MFCD06312295
  • Molecular Weight : 227.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart