PRIH93ED7A9F Princeton BioMolecular Research, Inc.

N-(2-piperidin-1-ylethyl)-1H-benzimidazol-2-amine

  • MDL number MFCD09259272
  • Molecular Weight : 244.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart