PRIH93ED7AA5 Princeton BioMolecular Research, Inc.

2-methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole-8-carboxylic acid

  • MDL number MFCD01181836
  • Molecular Weight : 230.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart