PRIH93ED7AB4 Princeton BioMolecular Research, Inc.

3-(carboxymethyl)-5-methyl-1H-indole-2-carboxylic acid

  • MDL number MFCD09259535
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart