PRIH93ED7AB7 Princeton BioMolecular Research, Inc.

5-Chloro-2-ethyl-1H-benzoimidazole

  • MDL number MFCD00995436
  • Cas Number : 34569-15-8
  • Molecular Weight : 180.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart