PRIH93ED7AC4 Princeton BioMolecular Research, Inc.

3-tert-butyl-2,3,4,9-tetrahydro-1H-carbazole

  • MDL number MFCD02942368
  • Cas Number : 22401-71-4
  • Molecular Weight : 227.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart