PRIH93ED7ACF Princeton BioMolecular Research, Inc.

2-(ethylthio)-5-nitro-1H-benzimidazole

  • MDL number MFCD00459120
  • Molecular Weight : 223.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart