PRIH93ED7ADF Princeton BioMolecular Research, Inc.

methyl (2-benzyl-1H-benzimidazol-1-yl)acetate

  • MDL number MFCD02859131
  • Molecular Weight : 280.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart