PRIH93ED7AE6 Princeton BioMolecular Research, Inc.

1-(4-(2-(1H-benzimidazol-1-yl)ethoxy)phenyl)ethanone

  • MDL number MFCD02736278
  • Molecular Weight : 280.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart