PRIH93ED7AEA Princeton BioMolecular Research, Inc.

(2-anilino-1H-benzimidazol-1-yl)acetic acid

  • MDL number MFCD08559426
  • Molecular Weight : 267.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart