PRIH93ED7AFA Princeton BioMolecular Research, Inc.

2-(2-(methylthio)-1H-benzimidazol-1-yl)acetamide

  • MDL number MFCD00612469
  • Molecular Weight : 221.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart