PRIH93ED7AFD Princeton BioMolecular Research, Inc.

9-(2-(2-methylphenoxy)ethyl)-9H-purin-6-amine

  • MDL number MFCD03928130
  • Molecular Weight : 269.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart