PRIH93ED7B10 Princeton BioMolecular Research, Inc.

8-bromo-2,4-dimethylquinoline

  • MDL number MFCD09754127
  • Cas Number : 1023812-38-5
  • Molecular Weight : 236.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart