PRIH93ED7B1E Princeton BioMolecular Research, Inc.

7,7-dimethyl-2-oxobicyclo[2.2.1]heptane-1-carbonitrile

  • MDL number MFCD09754232
  • Molecular Weight : 163.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart