PRIH93ED7B22 Princeton BioMolecular Research, Inc.

(2-(hydroxyimino)-1,7-dimethylbicyclo[2.2.1]hept-7-yl)acetonitrile

  • MDL number MFCD08447154
  • Molecular Weight : 192.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart