PRIH93ED7B29 Princeton BioMolecular Research, Inc.

6-bromo-4,5,5-trimethylbicyclo[2.1.1]hexane-1-carboxylic acid

  • MDL number MFCD03731361
  • Molecular Weight : 247.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart