PRIH93ED7B38 Princeton BioMolecular Research, Inc.

2-amino-2-methyl-2,3-dihydro-1H-isoindolium bromide

  • MDL number MFCD06496254
  • Molecular Weight : 229.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart