PRIH93ED7B46 Princeton BioMolecular Research, Inc.

5-chloro-2-((1-phenylethyl)amino)pyrimidine-4-carboxylic acid

  • MDL number MFCD09754276
  • Molecular Weight : 277.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart