PRIH93ED7B56 Princeton BioMolecular Research, Inc.

N-ethyl-N'-(2-methyl-5-nitrophenyl)thiourea

  • MDL number MFCD05871718
  • Molecular Weight : 239.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart