PRIH93ED7B5D Princeton BioMolecular Research, Inc.

N-ethyl-N'-(3-(trifluoromethyl)phenyl)thiourea

  • MDL number MFCD06113776
  • Molecular Weight : 248.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart