PRIH93ED7B5E Princeton BioMolecular Research, Inc.

N-(4-methoxyphenyl)-N'-methylthiourea

  • MDL number MFCD02320822
  • Molecular Weight : 196.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart