PRIH93ED7B7B Princeton BioMolecular Research, Inc.

1-methyl-1H-indole-2,3-dione 3-thiosemicarbazone

  • MDL number MFCD00044199
  • Molecular Weight : 234.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart